Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279056_m1
          See other ZFP941 GE Assays ›
          Gene Symbol:
          Zfp941
          Gene Name:
          zinc finger protein 941
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 140809677 - 140822178 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279056_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp941
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001180.2 NP_001001180.2 2 - 3 271 126
          GenBank mRNA AK045889.1 - 2 - 3 250 126
          AK141045.1 - 249 126
          BB622084.1 - 2 - 3 273 126
          BC066028.1 - 2 - 3 225 5702088 126
          BC068250.1 - 2 - 3 241 30018777 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004506.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp941

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062652 BC066028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062652 BC068250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062652 AK141045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062652 NM_001001180.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109479 BC066028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109479 NM_001001180.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151825 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141260 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154026 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109479 BC066028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109479 NM_001001180.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062652 BC066028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062652 BC068250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062652 AK141045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062652 NM_001001180.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25585087 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141260 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154026 - -
          Whole Mouse Genome G4122A A_52_P365970 BC066028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365970 BC068250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365970 NM_001001180.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105500 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154026 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141260 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05933063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05886218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05933058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05933051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25585035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06227194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06251907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06396305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05933052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05948183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05891672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05933056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25585083 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568983 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568987 - -
          Affymetrix HT MG-430 PM Array Plate 1439850_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458163_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568982 - -
          Mouse Genome 430 2.0 Array 1458163_PM_at - -
          Mouse Genome 430 2.0 Array 1439850_PM_at - -
          Mouse Genome 430 2.0 Array 1458163_at - -
          Mouse Genome 430 2.0 Array 1439850_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 407812_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 407812_at - -
          Affymetrix Mouse Clariom S Array PSR0700037841.mm.1 NM_001001180.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037841.mm.1 NM_001001180.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037841.mm.1 TC0700004506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037842.mm.1 NM_001001180.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037842.mm.1 NM_001001180.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037842.mm.1 TC0700004506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037840.mm.1 NM_001001180.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037840.mm.1 NM_001001180.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037840.mm.1 TC0700004506.mm.1 MTA TC
          Mouse Expression Set 430 1439850_at - -
          Mouse Expression Set 430 1458163_at - -
          Mouse Expression Set 430 1439850_PM_at - -
          Mouse Expression Set 430 1458163_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439850_at - -
          Affymetrix HT Mouse Genome 430B Array 1458163_at - -
          Affymetrix HT Mouse Genome 430B Array 1458163_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439850_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 817890 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162180 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162180 - -
          mouse-6 v1.1 (Illumina) GI_47564081-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162180 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162180 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_47564081-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline