Search
Search

H2-M9| Gene Symbol: H2-M9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008205.1 | NP_032231.1 | 1 - 2 | 78 | 70 | |
| GenBank mRNA | AF469488.1 | - | 1 - 2 | 78 | 70 | |
| AF539595.1 | - | 1 - 2 | 78 | 70 | ||
| AK080256.1 | - | 1 - 2 | 102 | 70 | ||
| BC106996.2 | - | 1 - 2 | 77 | 70 | ||
Set Membership: |
|