| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700019438.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019438.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019438.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019438.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019438.mm.1 |
TC1700002038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700019436.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019436.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019436.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019436.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019436.mm.1 |
TC1700002038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700019435.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019435.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019435.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019435.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019435.mm.1 |
TC1700002038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700019439.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019439.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019439.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019439.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019439.mm.1 |
TC1700002038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700019434.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019434.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019434.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019434.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019434.mm.1 |
TC1700002038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700019433.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019433.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019433.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019433.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019433.mm.1 |
TC1700002038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700019437.mm.1 |
NM_001033235.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019437.mm.1 |
NM_001033235.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019437.mm.1 |
NM_001172168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019437.mm.1 |
NM_001172168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700019437.mm.1 |
TC1700002038.mm.1 |
MTA TC |