Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280188_s1
          See other 4931431F19RIK GE Assays ›
          Gene Symbol:
          4931431F19Rik
          Gene Name:
          RIKEN cDNA 4931431F19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 104127913 - 104129823 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280188_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4931431F19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027634.1 NP_081910.1 1 1738 64
          GenBank mRNA AK016497.1 - 1 1739 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243454 NM_027634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243454 AK016497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243454 NM_027634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243454 AK016497.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172151 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243454 NM_027634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243454 AK016497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243454 NM_027634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243454 AK016497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243454 NM_027634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243454 AK016497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243454 NM_027634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243454 AK016497.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06140083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06140075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386719 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453516_PM_at - -
          Mouse Genome 430 2.0 Array 1453516_at - -
          Mouse Genome 430 2.0 Array 1453516_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70980_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70980_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700032842.mm.1 NM_027634.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032842.mm.1 NM_027634.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032842.mm.1 TC0700003927.mm.1 MTA TC
          Mouse Expression Set 430 1453516_at - -
          Mouse Expression Set 430 1453516_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453516_at - -
          Affymetrix HT Mouse Genome 430B Array 1453516_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 331903 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104280092 - -
          Illumina mouse-6 v1.0 expression beadchip 104280092 - -
          Illumina mouse-6 v1.0 expression beadchip 101230750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255459 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249345 - -
          mouse-6 v1.1 (Illumina) GI_38087826-S - -
          mouse-6 v1.1 (Illumina) scl30917.1.2_197-S - -
          Illumina Sentrix mouse-6.v1 bead array 104280092 - -
          Illumina Sentrix mouse-6.v1 bead array 101230750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104280092 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101230750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255459 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249345 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline