Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280303_s1
          See other OLFR646 GE Assays ›
          Gene Symbol:
          Olfr646
          Gene Name:
          olfactory receptor 646
          Gene Aliases:
          MOR33-2
          Chromosome Location:
          Chr.7: 104106281 - 104107219 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280303_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr646
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147056.1 NP_667267.1 1 896 64
          GenBank mRNA BC127980.1 - 1 900 40135216 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr646

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303851 NM_147056.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303851 BC127980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303851 NM_147056.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303851 BC127980.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171395 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303851 NM_147056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303851 BC127980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303851 NM_147056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303851 BC127980.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303851 NM_147056.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303851 BC127980.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303851 NM_147056.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303851 BC127980.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171395 - -
          Whole Mouse Genome G4122A A_51_P303851 NM_147056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303851 BC127980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303851 NM_147056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303851 BC127980.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134130 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06217601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06234481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06234482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06217607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555845 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555844 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 477457 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3290377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219024 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219024 - -
          mouse-6 v1.1 (Illumina) scl0259058.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0259058.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219024 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219024 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline