Search
Search

Olfr559| Gene Symbol: Olfr559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147112.1 | NP_667323.1 | 1 | 50 | 62 | |
| GenBank mRNA | BC119185.1 | - | 1 | 72 | 8733934 | 62 |
| BC120640.1 | - | 1 | 72 | 40129563 | 62 | |
Set Membership: |
|