Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280497_s1
          See other OLFR130 GE Assays ›
          Gene Symbol:
          Olfr130
          Gene Name:
          olfactory receptor 130
          Gene Aliases:
          MOR256-19
          Chromosome Location:
          Chr.17: 38067173 - 38068126 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280497_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr130
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146487.1 NP_666698.1 1 905 151
          GenBank mRNA BC128267.1 - 1 905 40043327 151

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr130

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999752 BC128267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999752 NM_146487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102603 BC128267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102603 NM_146487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999752 BC128267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999752 NM_146487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102603 BC128267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102603 NM_146487.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P20688 BC128267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P20688 NM_146487.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170982 - -
          Whole Mouse Genome G4122A A_52_P20688 BC128267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P20688 NM_146487.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131896 NM_207155.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131896 BC148220.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131896 BC148277.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13176392 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13471585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13471596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31171000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13213052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445173 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445172 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 334676 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2680059 - -
          Illumina mouse-6 v1.0 expression beadchip 2680059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711432 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711432 - -
          mouse-6 v1.1 (Illumina) scl0258480.1_158-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258480.1_158-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711432 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0