Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280891_s1
          See other OLFR506 GE Assays ›
          Gene Symbol:
          Olfr506
          Gene Name:
          olfactory receptor 506
          Gene Aliases:
          MOR204-23
          Chromosome Location:
          Chr.7: 108612309 - 108613253 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280891_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr506
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011871.1 NP_001011871.1 1 519 129
          GenBank mRNA BC134384.1 - 523 129
          BC147670.1 - 1 548 9056643 129
          BC147675.1 - 1 548 9056648 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr506

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045434 BC147670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045434 BC147675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045434 NM_001011871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045434 BC134384.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045434 BC147670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045434 BC147675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045434 NM_001011871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045434 BC134384.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403947 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05977114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05977108 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05977119 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05977125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05977122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05977113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403942 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556155 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556154 - -
          Affymetrix Mouse Clariom S Array PSR0700013283.mm.1 TC0700001600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013285.mm.1 TC0700001600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013284.mm.1 TC0700001600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013284.mm.1 NM_001011871.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013284.mm.1 NM_001011871.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 342045 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2813771 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2813771 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0