Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281113_s1
          See other OLFR1145 GE Assays ›
          Gene Symbol:
          Olfr1145
          Gene Name:
          olfactory receptor 1145
          Gene Aliases:
          MOR264-19
          Chromosome Location:
          Chr.2: 87809822 - 87810799 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281113_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1145
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146320.2 NP_666432.2 1 135 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1145

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372090 NM_146320.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372090 NM_146320.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293401 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372090 NM_146320.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372090 NM_146320.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P372090 NM_146320.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P372090 NM_146320.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407320 - -
          Whole Mouse Genome G4122A A_52_P372090 NM_146320.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372090 NM_146320.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112162 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01525268 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01999337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01525273 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01870832 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01870830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01256646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01256638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01686241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01431623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473567 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473566 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 623031 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6180131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617646 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617646 - -
          mouse-6 v1.1 (Illumina) scl4910.1.1_226-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4910.1.1_226-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2617646 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617646 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xmktn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0