Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281135_sH
          See other OLFR117 GE Assays ›
          Gene Symbol:
          Olfr117
          Gene Name:
          olfactory receptor 117
          Gene Aliases:
          MOR256-33
          Chromosome Location:
          Chr.17: 37659378 - 37660331 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281135_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr117
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207155.2 NP_997038.2 1 762 87
          GenBank mRNA BC148220.1 - 1 766 40135828 87
          BC148277.1 - 1 766 40135829 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr117

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131896 NM_207155.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131896 BC148220.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131896 BC148277.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156343 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131896 NM_207155.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131896 BC148220.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131896 BC148277.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169477 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156343 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13434014 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13318641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13434017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13318631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450903 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450902 - -
          Affymetrix Mouse Clariom S Array PSR1700019618.mm.1 NM_207155.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019618.mm.1 NM_207155.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019618.mm.1 TC1700002078.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 780398 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780398 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662165 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2662165 - -
          mouse-6 v1.1 (Illumina) scl0258263.1_166-S - -
          Illumina Sentrix mouse-6.v1 bead array 780398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258263.1_166-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662165 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline