Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281184_s1
          See other OLFR553 GE Assays ›
          Gene Symbol:
          Olfr553
          Gene Name:
          olfactory receptor 553
          Gene Aliases:
          MOR25-2
          Chromosome Location:
          Chr.7: 102614007 - 102614987 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281184_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr553
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207621.1 NP_997504.1 1 916 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr553

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123037 NM_207621.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31248 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149945 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130607 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131909 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123037 NM_207621.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149945 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130607 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119847 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130607 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149945 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06241807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06084578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06241805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06241798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06638273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06241796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06638273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06337668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06468961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06337664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379773 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566160 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566159 - -
          Affymetrix Mouse Clariom S Array PSR0700032647.mm.1 NM_207621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032647.mm.1 NM_207621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032647.mm.1 TC0700003864.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 321105 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6650619 - -
          Illumina mouse-6 v1.0 expression beadchip 6650619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603782 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603782 - -
          mouse-6 v1.1 (Illumina) scl0233578.1_325-S - -
          Illumina Sentrix mouse-6.v1 bead array 6650619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0233578.1_325-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2603782 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2603782 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0