Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281421_g1
          See other TSPO2 GE Assays ›
          Gene Symbol:
          Tspo2
          Gene Name:
          translocator protein 2
          Gene Aliases:
          2510027D20Rik, Bzrpl1
          Chromosome Location:
          Chr.17: 48444411 - 48451635 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281421_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspo2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027292.2 NP_081568.1 4 - 5 571 82
          GenBank mRNA AK011003.1 - 4 - 5 572 82
          BC104344.1 - 3 - 4 430 40049489 82
          BC104345.1 - 3 - 4 430 40049491 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002227.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tspo2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          cholesterol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161929 AK162634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161929 NM_027292.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161929 AK011003.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161929 AK162634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161929 NM_027292.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161929 AK011003.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41780 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156365 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161929 AK162634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161929 NM_027292.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161929 AK011003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161929 AK162634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161929 NM_027292.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161929 AK011003.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217547 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156365 - -
          Whole Mouse Genome G4122A A_51_P161929 AK162634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161929 NM_027292.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161929 AK011003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161929 AK162634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161929 NM_027292.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161929 AK011003.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108593 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13256808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13500533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13192316 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13135955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13500531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13256815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13371304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13500532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13500533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13075901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451672 - -
          Affymetrix HT MG-430 PM Array Plate 1453047_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451670 - -
          Mouse Genome 430 2.0 Array 1453047_PM_at - -
          Mouse Genome 430 2.0 Array 1453047_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70026_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70026_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700021057.mm.1 NM_027292.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021057.mm.1 NM_027292.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021057.mm.1 TC1700002227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021056.mm.1 NM_027292.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021056.mm.1 NM_027292.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021056.mm.1 TC1700002227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021059.mm.1 NM_027292.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021059.mm.1 NM_027292.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021059.mm.1 TC1700002227.mm.1 MTA TC
          Mouse Expression Set 430 1453047_PM_at - -
          Mouse Expression Set 430 1453047_at - -
          Murine Genome U74v2 Set 103512_at - -
          Murine Genome U74v2 Set 103512_PM_at - -
          Mouse Genome 430A 2.0 Array 1453047_PM_at - -
          Mouse Genome 430A 2.0 Array 1453047_at - -
          Affymetrix HT Mouse Genome 430A Array 1453047_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453047_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 368394 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4480113 - -
          Illumina mouse-6 v1.0 expression beadchip 4480113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2635718 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2635718 - -
          mouse-6 v1.1 (Illumina) scl49830.2.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49830.2.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2635718 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2635718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline