Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281466_s1
          See other 2310039H08RIK GE Assays ›
          Gene Symbol:
          2310039H08Rik
          Gene Name:
          RIKEN cDNA 2310039H08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 46772635 - 46773407 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281466_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310039H08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025966.3 NP_080242.1 1 707 101
          GenBank mRNA AK003321.1 - 1 520 101
          AK009696.1 - 1 530 101
          BC038674.1 - 1 707 1513865 101
          BU055991.1 - 1 502 101

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2310039H08Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208847 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 AK003321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 AK009696.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 BC038674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 BU055991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 NM_025966.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 AK003321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 AK009696.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 BC038674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 BU055991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214083 NM_025966.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23955 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120608 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 AK003321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 AK009696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 BC038674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 BU055991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 NM_025966.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 AK003321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 AK009696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 BC038674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 BU055991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214083 NM_025966.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208844 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120608 - -
          Whole Mouse Genome G4122A A_51_P214083 AK003321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 AK009696.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 BC038674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 BU055991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 NM_025966.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 AK003321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 AK009696.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 BC038674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 BU055991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214083 NM_025966.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121074 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125957 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13225573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13225566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13499458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13207925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13416630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13225560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445628 - -
          Affymetrix HT MG-430 PM Array Plate 1417348_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445627 - -
          Mouse Genome 430 2.0 Array 1417348_PM_at - -
          Mouse Genome 430 2.0 Array 1417348_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67101_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67101_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700008649.mm.1 NM_025966.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008649.mm.1 NM_025966.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008649.mm.1 TC1700000902.mm.1 MTA TC
          Mouse Expression Set 430 1417348_at - -
          Mouse Expression Set 430 1417348_PM_at - -
          Mouse Genome 430A 2.0 Array 1417348_PM_at - -
          Mouse Genome 430A 2.0 Array 1417348_at - -
          Affymetrix HT Mouse Genome 430A Array 1417348_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417348_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 683066 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3870446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619836 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619836 - -
          mouse-6 v1.1 (Illumina) scl50670.2.184_270-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50670.2.184_270-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2619836 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619836 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50670.2.184_270-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline