Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281482_m1
          See other RSPH9 GE Assays ›
          Gene Symbol:
          Rsph9
          Gene Name:
          radial spoke head 9 homolog (Chlamydomonas)
          Gene Aliases:
          1700027N10Rik
          Chromosome Location:
          Chr.17: 46122038 - 46144237 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281482_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsph9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029338.3 NP_083614.1 4 - 5 720 60
          GenBank mRNA AK006440.1 - 4 - 5 723 60
          AK165820.1 - 4 - 5 591 60
          BC019423.1 - 4 - 5 694 4922914 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002164.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsph9

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cilium movement
          axoneme assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 NM_029338.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 AK006440.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 AK165820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 BC019423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 NM_029338.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 AK006440.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 AK165820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435922 BC019423.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133689 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156970 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121323 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 NM_029338.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 AK006440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 AK165820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 BC019423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 NM_029338.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 AK006440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 AK165820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435922 BC019423.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31205339 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121323 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03324270 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156970 - -
          Whole Mouse Genome G4122A A_51_P435922 NM_029338.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 AK006440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 AK165820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 BC019423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 NM_029338.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 AK006440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 AK165820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435922 BC019423.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109992 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156970 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121323 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13319529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13202706 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13202700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13202702 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13202701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13319530 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13202696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13194311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13202705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13554957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13471513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13176677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13202707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13550291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13372760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13319531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13129969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451218 - -
          Affymetrix HT MG-430 PM Array Plate 1424763_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451213 - -
          Mouse Genome 430 2.0 Array 1424763_at - -
          Mouse Genome 430 2.0 Array 1457813_PM_at - -
          Mouse Genome 430 2.0 Array 1457813_at - -
          Mouse Genome 430 2.0 Array 1424763_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75564_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75564_at - -
          Affymetrix Mouse Clariom S Array PSR1700020316.mm.1 NM_029338.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020316.mm.1 NM_029338.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020316.mm.1 TC1700002164.mm.1 MTA TC
          Mouse Expression Set 430 1424763_at - -
          Mouse Expression Set 430 1424763_PM_at - -
          Mouse Genome 430A 2.0 Array 1424763_PM_at - -
          Mouse Genome 430A 2.0 Array 1424763_at - -
          Affymetrix HT Mouse Genome 430A Array 1424763_at - -
          Affymetrix HT Mouse Genome 430A Array 1424763_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 435760 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1090093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775220 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775220 - -
          mouse-6 v1.1 (Illumina) scl49876.5.1_70-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49876.5.1_70-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2935237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775220 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2935237 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49876.5.1_70-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline