Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281492_g1
          See other LRRC73 GE Assays ›
          Gene Symbol:
          Lrrc73
          Gene Name:
          leucine rich repeat containing 73
          Gene Aliases:
          Gm88
          Chromosome Location:
          Chr.17: 46253445 - 46257316 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281492_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001111142.1 NP_001104612.1 5 - 6 1063 68
          GenBank mRNA AK134769.1 - 3 - 4 2129 68
          BC147714.1 - 5 - 6 1038 9087473 68
          BC172136.1 - 5 - 6 999 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000886.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P734742 AK134769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P734742 NM_001111142.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P734742 BC147714.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P734742 AK134769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P734742 NM_001111142.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P734742 BC147714.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29904 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131334 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P734742 AK134769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P734742 NM_001111142.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P734742 BC147714.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P734742 AK134769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P734742 NM_001111142.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P734742 BC147714.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131334 - -
          Whole Mouse Genome G4122A A_52_P734742 AK134769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P734742 NM_001111142.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P734742 BC147714.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P734742 AK134769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P734742 NM_001111142.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P734742 BC147714.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31205963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13099570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13271709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13166252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13491373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13352746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13491379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31205990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445510 - -
          Affymetrix HT MG-430 PM Array Plate 1455755_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445507 - -
          Mouse Genome 430 2.0 Array 1455755_PM_at - -
          Mouse Genome 430 2.0 Array 1455755_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224813_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224813_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700008457.mm.1 NM_001111142.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008457.mm.1 NM_001111142.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008457.mm.1 TC1700000886.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008456.mm.1 NM_001111142.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008456.mm.1 NM_001111142.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008456.mm.1 TC1700000886.mm.1 MTA TC
          Mouse Expression Set 430 1455755_at - -
          Mouse Expression Set 430 1455755_PM_at - -
          Murine Genome U74v2 Set 130682_at - -
          Murine Genome U74v2 Set 130682_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455755_at - -
          Affymetrix HT Mouse Genome 430B Array 1455755_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 576743 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline