Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281591_m1
          See other NR5A1 GE Assays ›
          Gene Symbol:
          Nr5a1
          Gene Name:
          nuclear receptor subfamily 5, group A, member 1
          Gene Aliases:
          Ad4BP, ELP, ELP-3, Ftz-F1, Ftzf1, SF-1, SF1
          Chromosome Location:
          Chr.2: 38692656 - 38714560 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281591_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr5a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139051.3 NP_620639.1 1 - 2 191 77
          GenBank mRNA AK134378.1 - 1 - 2 192 77
          AK144376.1 - 1 - 2 152 77
          AK156911.1 - 1 - 2 159 77
          BC110476.1 - 1 - 2 134 40047261 77
          BC110477.1 - 1 - 2 134 40047262 77

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK134378.1, AK144376.1, AK156911.1, BC110476.1, BC110477.1, NM_139051.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr5a1

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          luteinization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          male gonad development
          tissue development
          multicellular organism aging
          reproductive process
          cell differentiation
          adrenal gland development
          hormone metabolic process
          steroid hormone mediated signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          maintenance of protein location in nucleus
          positive regulation of male gonad development
          negative regulation of female gonad development
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          protein binding
          phospholipid binding
          zinc ion binding
          lipid binding
          enzyme binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200029356.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029356.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029356.mm.1 TC0200003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029357.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029357.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029357.mm.1 TC0200003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029352.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029352.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029352.mm.1 TC0200003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029353.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029353.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029353.mm.1 TC0200003423.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0