Search
Search

Rftn1| Gene Symbol: Rftn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181397.2 | NP_852062.1 | 3 - 4 | 621 | 66 | |
| GenBank mRNA | AK082517.1 | - | 234 | 66 | ||
| AK087803.1 | - | 394 | 66 | |||
| AK152130.1 | - | 3 - 4 | 543 | 66 | ||
| AK153288.1 | - | 3 - 4 | 543 | 66 | ||
| AK155870.1 | - | 3 - 4 | 585 | 66 | ||
| AK170913.1 | - | 3 - 4 | 584 | 66 | ||
| AK172884.1 | - | 3 - 4 | 549 | 66 | ||
| BC052074.1 | - | 3 - 4 | 621 | 6401192 | 66 | |
Set Membership: |
|