Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281666_m1
          See other BTNL2 GE Assays ›
          Gene Symbol:
          Btnl2
          Gene Name:
          butyrophilin-like 2
          Gene Aliases:
          BTL-II, BTLN2, Btl2, Gm315, NG9
          Chromosome Location:
          Chr.17: 34354789 - 34369492 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281666_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Btnl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_079835.2 NP_524574.1 6 - 7 1336 69
          GenBank mRNA AK136521.1 - 6 - 7 1338 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000629.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Btnl2

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell proliferation
          negative regulation of T cell receptor signaling pathway
          positive regulation of interleukin-2 secretion
          receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995354 NM_079835.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995354 AK136521.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39727 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995354 NM_079835.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1995354 AK136521.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31153418 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03310333 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08765203 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03310309 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08765203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31153447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13428347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13522273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13280882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13241613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13108324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13055070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13426552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13297044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13380477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13319037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13179211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13496353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13483569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13216660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13319041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13108327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13380474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31153547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13319044 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444314 - -
          Affymetrix HT MG-430 PM Array Plate 1442199_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444312 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 547431_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 547431_at - -
          Affymetrix Mouse Clariom S Array PSR1700006075.mm.1 NM_079835.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006075.mm.1 NM_079835.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006075.mm.1 TC1700000629.mm.1 MTA TC
          Mouse Expression Set 430 1442199_at - -
          Mouse Expression Set 430 1442199_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 449525 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2530687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2779654 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2779654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline