| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016622.mm.1 |
NM_001080777.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016622.mm.1 |
NM_001284205.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016622.mm.1 |
NM_025585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016622.mm.1 |
TC1300002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016623.mm.1 |
NM_001080777.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016623.mm.1 |
NM_001284205.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016623.mm.1 |
NM_025585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016623.mm.1 |
TC1300002475.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016621.mm.1 |
NM_001080777.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016621.mm.1 |
NM_025585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016621.mm.1 |
TC1300002475.mm.1 |
MTA TC |