Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01283978_m1
          See other NR1I3 GE Assays ›
          Gene Symbol:
          Nr1i3
          Gene Name:
          nuclear receptor subfamily 1, group I, member 3
          Gene Aliases:
          AA209988, AI551208, CAR, CAR-beta, Care2, ESTM32, MB67
          Chromosome Location:
          Chr.1: 171213935 - 171218845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01283978_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr1i3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243062.1 NP_001229991.1 3 - 4 484 68
          NM_001243063.1 NP_001229992.1 3 - 4 484 68
          NM_009803.5 NP_033933.2 3 - 4 484 68
          GenBank mRNA AF009327.1 - 3 - 4 421 68
          AF009328.1 - 3 - 4 518 68
          AK133515.1 - 3 - 4 485 68
          AK149381.1 - 3 - 4 436 68
          BC089541.1 - 3 - 4 433 30315989 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001579.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr1i3

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          steroid hormone mediated signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          thyroid hormone receptor activity
          protein binding
          zinc ion binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 NM_001243062.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 NM_001243062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 NM_001243063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 NM_001243063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 NM_009803.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 NM_009803.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012929.mm.1 TC0100001579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 NM_001243062.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 NM_001243062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 NM_001243063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 NM_001243063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 NM_009803.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 NM_009803.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012931.mm.1 TC0100001579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 NM_001243062.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 NM_001243062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 NM_001243063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 NM_001243063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 NM_009803.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 NM_009803.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012932.mm.1 TC0100001579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 NM_001243062.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 NM_001243062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 NM_001243063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 NM_001243063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 NM_009803.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 NM_009803.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012933.mm.1 TC0100001579.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 916213 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline