Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01283995_m1
          See other ILDR2 GE Assays ›
          Gene Symbol:
          Ildr2
          Gene Name:
          immunoglobulin-like domain containing receptor 2
          Gene Aliases:
          2810478N18Rik, 3110063L10Rik, AI852300, D1Ertd471e, Dbsm1, ENSMUSG00000040612, Ll
          Chromosome Location:
          Chr.1: 166254094 - 166316832 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01283995_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ildr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164528.1 NP_001158000.1 7 - 8 1272 73
          GenBank mRNA FJ024494.1 - 1218 73
          FJ024495.1 - 7 - 8 1218 73
          FJ024496.1 - 6 - 7 1041 73
          FJ024497.1 - 4 - 5 837 73
          FJ024498.1 - 6 - 7 1161 73
          FJ024499.1 - 5 - 6 894 73
          FJ024500.1 - 7 - 8 1218 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001533.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          response to glucose
          insulin secretion
          cell differentiation
          pancreas development
          homeostasis of number of cells within a tissue

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024500.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 NM_001164528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024500.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 NM_001164528.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04342440 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00242195 - -
          Whole Mouse Genome G4122A A_51_P452533 NM_001164528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P452533 AK014234.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P452533 FJ024494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221294 G49232.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138858 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00242195 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351383 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351386 - -
          Affymetrix HT MG-430 PM Array Plate 1436293_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1436894_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436221_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351380 - -
          Affymetrix Mouse Gene 1.1 ST Array 10359814 - -
          Mouse Genome 430 2.0 Array 1436894_PM_at - -
          Mouse Genome 430 2.0 Array 1436894_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100039795_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100039795_at - -
          Affymetrix Mouse Clariom S Array PSR0100012640.mm.1 NM_001164528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012640.mm.1 NM_001164528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012640.mm.1 TC0100001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012639.mm.1 NM_001164528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012639.mm.1 NM_001164528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012639.mm.1 TC0100001533.mm.1 MTA TC
          Mouse Expression Set 430 1436894_PM_at - -
          Mouse Expression Set 430 1436894_at - -
          Murine Genome U74v2 Set 169845_f_at - -
          Murine Genome U74v2 Set 169845_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1436293_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1436293_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1436894_at - -
          Affymetrix HT Mouse Genome 430B Array 1436221_at - -
          Affymetrix HT Mouse Genome 430B Array 1436894_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436221_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 526904 - -
          ABI Mouse Genome Survey Microarray 561161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline