Search
Search
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293313.1 | NP_001280242.1 | 5 - 6 | 637 | 59 | |
| NM_138314.4 | NP_612187.2 | 5 - 6 | 640 | 59 | ||
| NM_178071.6 | NP_835172.1 | 5 - 6 | 637 | 59 | ||
| GenBank mRNA | AF202048.1 | - | 4 - 5 | 541 | 59 | |
| AK044962.1 | - | 5 - 6 | 611 | 59 | ||
| AK085222.1 | - | 5 - 6 | 585 | 59 | ||
| AK133221.1 | - | 1108 | 59 | |||
| AK144915.1 | - | 5 - 6 | 641 | 59 | ||
| AK164652.1 | - | 5 - 6 | 585 | 59 | ||
| BC038021.1 | - | 6 - 7 | 667 | 3963844 | 59 | |
| BC049225.1 | - | 4 - 5 | 568 | 3483725 | 59 | |
Set Membership: |
|