Search
Search
| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137521.1 | - | 1 - 2 | 289 | 124 | |
| AK137537.1 | - | 1 - 2 | 266 | 124 | ||
| AK139240.1 | - | 1 - 2 | 283 | 124 | ||
| BY722729.1 | - | 1 - 2 | 263 | 124 | ||
| U52925.1 | - | 1 - 2 | 164 | 124 | ||
Set Membership: |
|