Search
Search

Trmt1l| Gene Symbol: Trmt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026876.3 | NP_081152.2 | 9 - 10 | 1535 | 69 | |
| GenBank mRNA | AK146088.1 | - | 9 - 10 | 1536 | 69 | |
| BC022682.1 | - | 4 - 5 | 571 | 69 | ||
| BC037478.1 | - | 5 - 6 | 715 | 69 | ||
| BC064024.1 | - | 9 - 10 | 1333 | 69 | ||
| BC110425.1 | - | 9 - 10 | 1518 | 5369595 | 69 | |
| BC132299.1 | - | 9 - 10 | 1393 | 40130576 | 69 | |
Set Membership: |
|