Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01285112_m1
          See other LCT GE Assays ›
          Gene Symbol:
          Lct
          Gene Name:
          lactase
          Gene Aliases:
          Gm100, LAC, LPH, Lphl
          Chromosome Location:
          Chr.1: 128284764 - 128328318 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01285112_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lct
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081078.1 NP_001074547.1 14 - 15 5134 61
          GenBank mRNA AK158042.1 - 8 - 9 3236 61
          AY751548.1 - 2 - 3 235 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003025.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          response to nutrient
          response to organic substance
          response to iron(II) ion
          response to nickel cation
          response to lead ion
          response to drug
          response to ethanol
          lactase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P164797 NM_001081078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P164797 NM_001081078.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916485 AF106967.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916485 AK080467.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916485 BC005677.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916485 BC021149.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916485 BC095955.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916485 NM_018772.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73129 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P164797 NM_001081078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P164797 NM_001081078.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00181270 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04257331 - -
          Whole Mouse Genome G4122A A_52_P164797 NM_001081078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P164797 NM_001081078.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04257331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20599896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00934651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00858535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00372442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00939407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20599840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10357432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357422 - -
          Affymetrix HT MG-430 PM Array Plate 1439479_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10357418 - -
          Mouse Genome 430 2.0 Array 1439479_PM_at - -
          Mouse Genome 430 2.0 Array 1439479_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226413_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226413_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100024226.mm.1 NM_001081078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024226.mm.1 TC0100003025.mm.1 MTA TC
          Mouse Expression Set 430 1439479_PM_at - -
          Mouse Expression Set 430 1439479_at - -
          Murine Genome U74v2 Set 104018_PM_at - -
          Murine Genome U74v2 Set 104018_at - -
          Mouse Genome 430A 2.0 Array 1439479_at - -
          Mouse Genome 430A 2.0 Array 1439479_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439479_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439479_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490831 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224973 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline