| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100023812.mm.1 |
NM_001115074.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023812.mm.1 |
NM_001115074.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023812.mm.1 |
TC0100002961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023813.mm.1 |
NM_001115074.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023813.mm.1 |
NM_001115074.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023813.mm.1 |
TC0100002961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023811.mm.1 |
NM_001115074.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023811.mm.1 |
NM_001115074.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023811.mm.1 |
TC0100002961.mm.1 |
MTA TC |