| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016134.mm.1 |
NM_001163627.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016134.mm.1 |
NM_001163627.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016134.mm.1 |
TC1300002422.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016133.mm.1 |
NM_001163627.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016133.mm.1 |
NM_001163627.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016133.mm.1 |
TC1300002422.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016135.mm.1 |
NM_001163627.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016135.mm.1 |
NM_001163627.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016135.mm.1 |
TC1300002422.mm.1 |
MTA TC |