Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01286607_s1
          See other OLFR51 GE Assays ›
          Gene Symbol:
          Olfr51
          Gene Name:
          olfactory receptor 51
          Gene Aliases:
          ID7, MOR129-3
          Chromosome Location:
          Chr.11: 51006974 - 51007897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01286607_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr51
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146909.1 NP_667120.1 1 78 84
          GenBank mRNA BC111814.1 - 1 77 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000557.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr51

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139690 NM_146909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139690 AF102525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139690 BC111814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139690 NM_146909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139690 AF102525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139690 BC111814.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152422 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139690 NM_146909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139690 AF102525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139690 BC111814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139690 NM_146909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139690 AF102525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139690 BC111814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139690 NM_146909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139690 AF102525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139690 BC111814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139690 NM_146909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139690 AF102525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139690 BC111814.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791805 - -
          Whole Mouse Genome G4122A A_51_P139690 NM_146909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139690 AF102525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139690 BC111814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139690 NM_146909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139690 AF102525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139690 BC111814.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139183 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09726223 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09617852 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09726215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09476624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09387673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09025853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09726221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09476625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09726225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09440372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09021149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09126278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09491583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09105364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09390484 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375794 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375791 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375793 - -
          Affymetrix Mouse Clariom S Array PSR1100004301.mm.1 NM_146909.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004301.mm.1 NM_146909.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004301.mm.1 TC1100000557.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 424512 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103710390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233298 - -
          mouse-6 v1.1 (Illumina) scl41603.1.190_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 103710390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103710390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2985602 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233298 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2985602 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0