Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01287179_m1
          See other KCNJ13 GE Assays ›
          Gene Symbol:
          Kcnj13
          Gene Name:
          potassium inwardly-rectifying channel, subfamily J, member 13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 87386284 - 87394729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01287179_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnj13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110227.1 NP_001103697.1 2 - 3 596 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002762.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Ion Channel 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          potassium ion transport
          potassium ion import
          regulation of ion transmembrane transport
          inward rectifier potassium channel activity
          voltage-gated ion channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066878 NM_001110227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081050 NM_001110227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066878 NM_001110227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081050 NM_001110227.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20417426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00066971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00086331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00528638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00827565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00654188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00922294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00681266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00553786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00908911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00553797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00086324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00553789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20417493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356404 - -
          Affymetrix HT MG-430 PM Array Plate 1456418_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10356403 - -
          Mouse Genome 430 2.0 Array 1456418_at - -
          Mouse Genome 430 2.0 Array 1456418_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100040591_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100040591_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100022289.mm.1 NM_001110227.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022289.mm.1 TC0100002762.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022294.mm.1 NM_001110227.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022294.mm.1 TC0100002762.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022288.mm.1 NM_001110227.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022288.mm.1 TC0100002762.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022290.mm.1 NM_001110227.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022290.mm.1 TC0100002762.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022292.mm.1 NM_001110227.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022292.mm.1 TC0100002762.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022291.mm.1 NM_001110227.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022291.mm.1 TC0100002762.mm.1 MTA TC
          Mouse Expression Set 430 1456418_PM_at - -
          Mouse Expression Set 430 1456418_at - -
          Affymetrix HT Mouse Genome 430B Array 1456418_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456418_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 318548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline