Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288934_m1
          See other BC053393 GE Assays ›
          Gene Symbol:
          BC053393
          Gene Name:
          cDNA sequence BC053393
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 46571513 - 46589232 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC053393
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025435.3 NP_001020606.2 4 - 5 661 110
          GenBank mRNA AK140690.1 - 4 - 5 516 110
          AK166706.1 - 4 - 5 538 110
          AK166838.1 - 4 - 5 538 110
          BC053393.1 - 2845 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000480.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BC053393

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092354 AK166838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092354 BC052651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092354 AK140690.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092354 AK166706.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092354 NM_001025435.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77580 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132428 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128080 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092354 AK166838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092354 BC052651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092354 AK140690.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092354 AK166706.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092354 NM_001025435.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27768597 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132428 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07329315 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07329284 - -
          Whole Mouse Genome G4122A A_52_P682557 AK166838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P682557 BC052651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P682557 BC053393.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P682557 AK140690.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P682557 AK166706.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128080 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09691570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09691574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09691565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27768720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09464188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09738269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09086885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09167795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09086883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09687595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09293465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27768576 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375456 - -
          Affymetrix HT MG-430 PM Array Plate 1447604_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10375454 - -
          Mouse Genome 430 2.0 Array 1447604_PM_at - -
          Mouse Genome 430 2.0 Array 1447604_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 407814_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 407814_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100003677.mm.1 NM_001025435.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003677.mm.1 NM_001025435.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003677.mm.1 TC1100000480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100003678.mm.1 NM_001025435.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003678.mm.1 NM_001025435.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003678.mm.1 TC1100000480.mm.1 MTA TC
          Mouse Expression Set 430 1447604_PM_at - -
          Mouse Expression Set 430 1447604_at - -
          Murine Genome U74v2 Set 103472_at - -
          Murine Genome U74v2 Set 103472_PM_at - -
          Mouse Genome 430A 2.0 Array 1447604_at - -
          Mouse Genome 430A 2.0 Array 1447604_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1447604_at - -
          Affymetrix HT Mouse Genome 430A Array 1447604_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 457174 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161532 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161532 - -
          mouse-6 v1.1 (Illumina) GI_70887608-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161532 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161532 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline