Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288959_m1
          See other THG1L GE Assays ›
          Gene Symbol:
          Thg1l
          Gene Name:
          tRNA-histidine guanylyltransferase 1-like (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 45945305 - 45955503 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288959_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thg1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080969.3 NP_001074438.1 5 - 6 773 62
          NM_001290737.1 NP_001277666.1 6 - 7 716 62
          GenBank mRNA AK010876.1 - 5 - 6 765 62
          AK015467.1 - 5 - 6 567 62
          AK016076.1 - 6 - 7 717 62
          AK017532.1 - 5 - 6 756 62
          BC092541.1 - 5 - 6 753 30280958 62
          BC115560.1 - 5 - 6 761 40101901 62
          BC115561.1 - 5 - 6 761 40101902 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002593.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA modification
          tRNA processing
          protein homotetramerization
          nucleotide binding
          magnesium ion binding
          ATP binding
          GTP binding
          tRNA guanylyltransferase activity
          transferase activity
          nucleotidyltransferase activity
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27765167 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27765169 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 BC115560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 BC115561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 AK010876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 AK015467.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 AK016076.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 BC115560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 BC115561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 AK010876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 AK015467.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204646 AK016076.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316311 AK012620.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316311 NM_007991.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58695 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139944 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 BC115560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 BC115561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 AK010876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 AK015467.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 AK016076.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 BC115560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 BC115561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 AK010876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 AK015467.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204646 AK016076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 BC115560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 BC115561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 AK010876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 AK015467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 AK016076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 BC115560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 BC115561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 AK010876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 AK015467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204646 AK016076.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09275308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09079238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09275310 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09275306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09275297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27765167 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27765169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27765158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09116563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09537681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09004689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09256193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09507390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27765129 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385377 - -
          Affymetrix HT MG-430 PM Array Plate 1432393_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1453268_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385375 - -
          Mouse Genome 430 2.0 Array 1453268_PM_at - -
          Mouse Genome 430 2.0 Array 1453268_at - -
          Mouse Genome 430 2.0 Array 1432393_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432393_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66628_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66628_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100023880.mm.1 TC1100002593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023881.mm.1 NM_001080969.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023881.mm.1 TC1100002593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023879.mm.1 TC1100002593.mm.1 MTA TC
          Mouse Expression Set 430 1453268_at - -
          Mouse Expression Set 430 1453268_PM_at - -
          Murine Genome U74v2 Set 168654_PM_at - -
          Murine Genome U74v2 Set 168654_at - -
          Affymetrix HT Mouse Genome 430B Array 1453268_at - -
          Affymetrix HT Mouse Genome 430B Array 1432393_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1432393_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1453268_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 877508 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227658 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227658 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2979430 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2979430 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline