Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01289097_m1
          See other NOVA1 GE Assays ›
          Gene Symbol:
          Nova1
          Gene Name:
          neuro-oncological ventral antigen 1
          Gene Aliases:
          9430099M15Rik, G630039L02, Nova-1
          Chromosome Location:
          Chr.12: 46694517 - 46819194 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01289097_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nova1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021361.1 NP_067336.1 4 - 5 539 70
          GenBank mRNA AF232828.1 - 4 - 5 473 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001692.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          nervous system development
          regulation of RNA metabolic process
          nucleic acid binding
          RNA binding
          mRNA binding
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174576 AK034094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174576 AK035201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174576 NM_021361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P204629 AK047565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P204629 NM_021361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P204629 AF232828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P204629 AK047565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P204629 NM_021361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P204629 AF232828.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894817 BC008262.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894817 BC051207.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832370 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82350 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136401 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147044 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131274 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174576 AK034094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174576 AK035201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174576 NM_021361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P204629 AK047565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P204629 NM_021361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P204629 AF232828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P204629 AK047565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P204629 NM_021361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P204629 AF232828.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610147 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610176 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147044 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610249 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02508142 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131274 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136401 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610090 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02508175 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610159 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610040 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07610111 - -
          Whole Mouse Genome G4122A A_52_P204629 AK047565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204629 NM_021361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204629 AF232828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204629 AK047565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204629 NM_021361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204629 AF232828.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131274 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02508142 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136401 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147044 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07610131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09983196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09895343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10327297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10419722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10329600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09822891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10122640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10329602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28409648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10233828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28410356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10400159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400160 - -
          Affymetrix HT MG-430 PM Array Plate 1426938_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452245_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10400157 - -
          Affymetrix Mouse Gene 1.1 ST Array 10400155 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 664883_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 664883_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200011698.mm.1 NM_021361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011698.mm.1 NM_021361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011698.mm.1 TC1200001692.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1452245_at - -
          Affymetrix HT Mouse Genome 430A Array 1452245_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426938_at - -
          Affymetrix HT Mouse Genome 430A Array 1426938_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617638 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105270685 - -
          MouseWG-6 v2 Expression BeadChip 102230324 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225424 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241644 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2535371 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0