Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164056.1 | NP_001157528.1 | 6 - 7 | 795 | 101 | |
NM_008875.4 | NP_032901.2 | 5 - 6 | 740 | 101 | ||
GenBank mRNA | BC068144.1 | - | 5 - 6 | 597 | 30532470 | 101 |
U87868.1 | - | 6 - 7 | 778 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |