Search
Search

Ccdc125| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168386.2 | NP_001161858.1 | 8 - 9 | 1196 | 69 | |
| NM_183115.5 | NP_898938.4 | 9 - 10 | 1274 | 69 | ||
| GenBank mRNA | AK017973.1 | - | 8 - 9 | 959 | 69 | |
| BC085249.1 | - | 9 - 10 | 992 | 30726150 | 69 | |
Set Membership: |
|