| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100005012.mm.1 |
NM_029105.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100005012.mm.1 |
TC1100000622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100005010.mm.1 |
TC1100000622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100005013.mm.1 |
NM_029105.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100005013.mm.1 |
TC1100000622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100005011.mm.1 |
NM_029105.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100005011.mm.1 |
TC1100000622.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100005009.mm.1 |
NM_029105.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100005009.mm.1 |
TC1100000622.mm.1 |
MTA TC |