Search
Search
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016696.4 | NP_057905.1 | 4 - 5 | 1123 | 108 | |
| GenBank mRNA | AF185613.1 | - | 4 - 5 | 1126 | 108 | |
| AK031768.1 | - | 91 | 108 | |||
| AK154584.1 | - | 2 - 3 | 257 | 108 | ||
| AK154899.1 | - | 4 - 5 | 1166 | 108 | ||
| AK196506.1 | - | 1 - 2 | 93 | 108 | ||
| BC062902.1 | - | 4 - 5 | 952 | 6810413 | 108 | |
Set Membership: |
|