Search
Search

Pm20d1| Gene Symbol: Pm20d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178079.3 | NP_835180.2 | 2 - 3 | 280 | 61 | |
| GenBank mRNA | AK028883.1 | - | 2 - 3 | 277 | 61 | |
| AK128969.1 | - | 2 - 3 | 286 | 61 | ||
| AK143953.1 | - | 2 - 3 | 275 | 61 | ||
| BC120725.1 | - | 2 - 3 | 280 | 40129648 | 61 | |
| BC125395.1 | - | 2 - 3 | 280 | 40129910 | 61 | |
Set Membership: |
|