Search
Search

Dis3l2| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172157.1 | NP_001165628.1 | 9 - 10 | 1156 | 63 | |
| NM_153530.2 | NP_705758.1 | 8 - 9 | 1114 | 63 | ||
| GenBank mRNA | AK029652.1 | - | 9 - 10 | 1170 | 63 | |
| AK031180.1 | - | 9 - 10 | 1139 | 63 | ||
| AK040613.1 | - | 10 - 11 | 1211 | 63 | ||
| AK214096.1 | - | 1 - 2 | 47 | 63 | ||
| BC036177.1 | - | 8 - 9 | 1075 | 5004341 | 63 | |
Set Membership: |
|