Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291309_m1
          See other PIK3C2B GE Assays ›
          Gene Symbol:
          Pik3c2b
          Gene Name:
          phosphoinositide-3-kinase, class 2, beta polypeptide
          Gene Aliases:
          C330011J12Rik, Gm199, PI3K-C2beta, PI3KC2beta
          Chromosome Location:
          Chr.1: 133045672 - 133108688 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291309_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pik3c2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099276.2 NP_001092746.2 20 - 21 3655 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001201.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          protein phosphorylation
          inflammatory response
          phosphatidylinositol 3-kinase signaling
          platelet activation
          phosphatidylinositol-3-phosphate biosynthetic process
          protein kinase B signaling
          innate immune response
          lipid phosphorylation
          phosphatidylinositol phosphorylation
          cell chemotaxis
          lipid kinase activity
          protein binding
          1-phosphatidylinositol-3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625252 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625253 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625255 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168544 NM_001099276.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P186487 AK049181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P186487 AK049181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2164644 NM_001099276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P186487 AK049181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P186487 AK049181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168544 NM_001099276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2164644 NM_001099276.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04268442 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00189094 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04268483 - -
          Whole Mouse Genome G4122A A_51_P186487 AK049181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P186487 AK049181.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625253 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625252 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20625254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00722833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00338936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00036178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00125344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00085562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01040774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01123506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00295319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00949184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00118922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00085553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00197263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00785117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00338937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01123511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00175470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00085556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00921700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00530432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00830326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00295317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00263418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01123508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00785116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00949177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00924718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00138781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01123512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00125354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00118926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00316932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00295315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00175469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00295309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00602399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00085561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00260931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00085554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00036170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01040772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00125360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00175468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00109969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00242616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01103977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00379099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00125347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20625242 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10349836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349838 - -
          Affymetrix Mouse Gene 1.1 ST Array 10349834 - -
          Affymetrix Mouse Clariom S Array PSR0100009794.mm.1 NM_001099276.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009794.mm.1 NM_001099276.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009794.mm.1 TC0100001201.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 302259 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244741 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2526530 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244741 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0