Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291968_g1
          See other GNB2L1 GE Assays ›
          Gene Symbol:
          Gnb2l1
          Gene Name:
          guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1
          Gene Aliases:
          AL033335, GB-like, Gnb2-rs1, Rack1, p205
          Chromosome Location:
          Chr.11: 48800360 - 48806241 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291968_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnb2l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008143.3 NP_032169.1 2 - 3 387 57
          GenBank mRNA AK002493.1 - 2 - 3 376 57
          AK012242.1 - 1 - 2 174 57
          AK017772.1 - 2 - 3 388 57
          AK051226.1 - 2 - 3 388 57
          AK159797.1 - 2 - 3 388 57
          AK160528.1 - 2 - 3 358 57
          AK160739.1 - 2 - 3 385 57
          AK161234.1 - 2 - 3 388 57
          AK166786.1 - 2 - 3 390 57
          AK166800.1 - 2 - 3 391 57
          AK166889.1 - 1 - 2 28 57
          AK166945.1 - 2 - 3 388 57
          AK168086.1 - 2 - 3 388 57
          AK168349.1 - 2 - 3 419 57
          AK169612.1 - 2 - 3 388 57
          BC020322.1 - 2 - 3 364 57
          BC046760.1 - 2 - 3 372 5693613 57
          D29802.1 - 2 - 3 381 57
          X75313.1 - 2 - 3 387 57

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnb2l1

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          positive regulation of protein phosphorylation
          regulation of translation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell cycle
          multicellular organism development
          gastrulation
          protein localization
          negative regulation of gene expression
          negative regulation of translation
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of cAMP catabolic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein homooligomerization
          negative regulation of peptidyl-serine phosphorylation
          intracellular signal transduction
          regulation of growth
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          rhythmic process
          negative regulation of phagocytosis
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          positive regulation of mitochondrial depolarization
          negative regulation of protein tyrosine kinase activity
          cellular response to glucose stimulus
          regulation of establishment of protein localization to plasma membrane
          negative regulation of hydrogen peroxide-induced neuron death
          positive regulation of intrinsic apoptotic signaling pathway
          receptor activity
          protein kinase C binding
          protein binding
          ion channel inhibitor activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          enzyme binding
          protein phosphatase binding
          protein tyrosine kinase inhibitor activity
          receptor tyrosine kinase binding
          signaling adaptor activity
          SH2 domain binding
          protein homodimerization activity
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100003730.mm.1 NM_008143.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003730.mm.1 NM_008143.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003730.mm.1 TC1100000493.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline