Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292181_g1
          See other 2310009B15RIK GE Assays ›
          Gene Symbol:
          2310009B15Rik
          Gene Name:
          RIKEN cDNA 2310009B15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 138851979 - 138856854 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292181_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310009B15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081226.1 NP_001074695.1 2 - 3 377 105
          GenBank mRNA AK009246.1 - 2 - 3 550 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P171724 NM_001081226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P171724 AK009246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P171724 NM_001081226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P171724 AK009246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388201 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084338 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P171724 NM_001081226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P171724 AK009246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P171724 NM_001081226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P171724 AK009246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P171724 NM_001081226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P171724 AK009246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P171724 NM_001081226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P171724 AK009246.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20659644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00546555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00202709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00394848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00152315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00202713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00546556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00152317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00202721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00035817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00546554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00802240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00202710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00394854 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608700 - -
          Affymetrix HT MG-430 PM Array Plate 1435776_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455761_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608700 - -
          Mouse Genome 430 2.0 Array 1435776_PM_at - -
          Mouse Genome 430 2.0 Array 1435776_at - -
          Mouse Genome 430 2.0 Array 1455761_PM_at - -
          Mouse Genome 430 2.0 Array 1455761_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69549_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69549_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100025766.mm.1 NM_001081226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025766.mm.1 NM_001081226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025766.mm.1 TC0100003176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025767.mm.1 NM_001081226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025767.mm.1 NM_001081226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025767.mm.1 TC0100003176.mm.1 MTA TC
          Mouse Expression Set 430 1435776_PM_at - -
          Mouse Expression Set 430 1455761_at - -
          Mouse Expression Set 430 1435776_at - -
          Mouse Expression Set 430 1455761_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435776_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455761_at - -
          Affymetrix HT Mouse Genome 430B Array 1435776_at - -
          Affymetrix HT Mouse Genome 430B Array 1455761_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580194 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229684 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline