Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292878_s1
          See other ALDOART2 GE Assays ›
          Gene Symbol:
          Aldoart2
          Gene Name:
          aldolase 1 A, retrogene 2
          Gene Aliases:
          4933425L11Rik, Aldo1-ps1, Aldoa-ps1
          Chromosome Location:
          Chr.12: 55565205 - 55566896 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292878_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldoart2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277340.1 NP_001264269.1 1 1582 86
          GenBank mRNA AK016920.1 - 1 1549 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063568 AK016920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063568 NR_003959.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063568 AK006425.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106130 - -
          Whole Mouse Genome G4122A A_51_P288903 AK016920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P288903 NR_003959.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P288903 AK006425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241612 AK016920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241612 EF662062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241612 NR_003959.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241612 AK006425.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10430014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09823435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09823432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10247540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10165536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10114458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09774498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10114460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10114461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10373179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28452508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395826 - -
          Affymetrix HT MG-430 PM Array Plate 1423498_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395825 - -
          Mouse Genome 430 2.0 Array 1423498_at - -
          Mouse Genome 430 2.0 Array 1423498_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200003260.mm.1 NM_001277340.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003260.mm.1 TC1200000471.mm.1 MTA TC
          Mouse Expression Set 430 1423498_PM_at - -
          Mouse Expression Set 430 1423498_at - -
          Murine Genome U74v2 Set 103613_PM_at - -
          Murine Genome U74v2 Set 103613_at - -
          Mouse Genome 430A 2.0 Array 1423498_PM_at - -
          Mouse Genome 430A 2.0 Array 1423498_at - -
          Affymetrix HT Mouse Genome 430A Array 1423498_at - -
          Affymetrix HT Mouse Genome 430A Array 1423498_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 817786 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0