Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011158.3 | NP_035288.2 | 1 - 2 | 499 | 56 | |
GenBank mRNA | AK011647.1 | - | 1 - 2 | 457 | 56 | |
AK041013.1 | - | 1 - 2 | 473 | 56 | ||
AK135621.1 | - | 1 - 2 | 501 | 56 | ||
AK138963.1 | - | 1 - 2 | 465 | 56 | ||
BC048710.1 | - | 1 - 2 | 455 | 6506827 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |