Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01294546_s1
          See other 4930402K13RIK GE Assays ›
          Gene Symbol:
          4930402K13Rik
          Gene Name:
          RIKEN cDNA 4930402K13 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 9104562 - 9106342 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01294546_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930402K13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270700.1 NP_001257629.1 1 1658 102
          GenBank mRNA AK015052.1 - 1 1659 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338510 AK015052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338510 AK015052.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155914 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338510 AK015052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338510 AK015052.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155914 - -
          Whole Mouse Genome G4122A A_51_P338510 AK015052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338510 AK015052.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113095 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14658744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14978959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245319 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453884_PM_at - -
          Mouse Genome 430 2.0 Array 1453884_at - -
          Mouse Genome 430 2.0 Array 1453884_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75775_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75775_PM_at - -
          Mouse Expression Set 430 1453884_PM_at - -
          Mouse Expression Set 430 1453884_at - -
          Murine Genome U74v2 Set 168785_at - -
          Murine Genome U74v2 Set 168785_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453884_at - -
          Affymetrix HT Mouse Genome 430B Array 1453884_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 686053 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106900528 - -
          MouseWG-6 v2 Expression BeadChip 101340086 - -
          Illumina mouse-6 v1.0 expression beadchip 106900528 - -
          Illumina mouse-6 v1.0 expression beadchip 101340086 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2767852 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2524573 - -
          mouse-6 v1.1 (Illumina) GI_38086064-S - -
          mouse-6 v1.1 (Illumina) scl55052.1.2_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 101340086 - -
          Illumina Sentrix mouse-6.v1 bead array 106900528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340086 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106900528 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2524573 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2767852 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0