Search
Search

Sec24a| Gene Symbol: Sec24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290785.1 | NP_001277714.1 | 13 - 14 | 2631 | 62 | |
| NM_175255.3 | NP_780464.2 | 13 - 14 | 2634 | 62 | ||
| GenBank mRNA | AK029139.1 | - | 13 - 14 | 2387 | 62 | |
| AK155182.1 | - | 13 - 14 | 2420 | 62 | ||
| BC117905.1 | - | 13 - 14 | 1898 | 62 | ||
Set Membership: |
|