Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295216_s1
          See other ANG5 GE Assays ›
          Gene Symbol:
          Ang5
          Gene Name:
          angiogenin, ribonuclease A family, member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 43957069 - 43963080 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295216_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ang5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007448.3 NP_031474.2 2 246 123

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ang5

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA phosphodiester bond hydrolysis
          molecular_function
          endonuclease activity
          ribonuclease activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134474 NM_001123394.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134474 U72672.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10414260 - -
          Mouse Genome 430 2.0 Array 1421791_PM_at - -
          Mouse Genome 430 2.0 Array 1421791_at - -
          Mouse Expression Set 430 1421791_at - -
          Mouse Expression Set 430 1421791_PM_at - -
          Murine Genome U74v2 Set 95228_PM_f_at - -
          Murine Genome U74v2 Set 95228_f_at - -
          Mouse Genome 430A 2.0 Array 1421791_PM_at - -
          Mouse Genome 430A 2.0 Array 1421791_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4570528 - -
          Illumina mouse-6 v1.0 expression beadchip 5290301 - -
          Illumina mouse-6 v1.0 expression beadchip 4570528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769355 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744316 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744316 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769355 - -
          mouse-6 v1.1 (Illumina) scl011730.3_53-S - -
          mouse-6 v1.1 (Illumina) scl0219033.2_317-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570528 - -
          Illumina Sentrix mouse-6.v1 bead array 5290301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0219033.2_317-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl011730.3_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290301 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570528 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744316 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769355 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0