Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295270_m1
          See other PCP4L1 GE Assays ›
          Gene Symbol:
          Pcp4l1
          Gene Name:
          Purkinje cell protein 4-like 1
          Gene Aliases:
          1500036F01Rik
          Chromosome Location:
          Chr.1: 171173264 - 171196268 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295270_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcp4l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025557.1 NP_079833.1 2 - 3 315 63
          GenBank mRNA AK002772.1 - 2 - 3 286 63
          AK032164.1 - 2 - 3 285 63
          AK075738.1 - 2 - 3 285 63
          AY304481.1 - 2 - 3 286 63
          BC116208.1 - 2 - 3 345 63
          BC126965.1 - 2 - 3 345 63
          BY238909.1 - 2 - 3 315 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003531.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983754 AK075738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983754 AY304481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983754 NM_025557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983754 AK002772.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156526 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167201 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140829 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983754 AK075738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983754 AY304481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983754 NM_025557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983754 AK002772.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00250509 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140829 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167201 - -
          Whole Mouse Genome G4122A A_51_P361150 AK075738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361150 AY304481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361150 NM_025557.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361150 AK002772.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113340 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167201 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00395297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00814306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00574728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00225655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00246076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00536279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00574737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00289132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01038162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01049338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00442028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00395294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00640160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00574730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00224937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00137811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826552 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360057 - -
          Affymetrix HT MG-430 PM Array Plate 1452913_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360053 - -
          Mouse Genome 430 2.0 Array 1452913_PM_at - -
          Mouse Genome 430 2.0 Array 1452913_at - -
          Mouse Genome 430 2.0 Array 1444791_at - -
          Mouse Genome 430 2.0 Array 1444791_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66425_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66425_at - -
          Affymetrix Mouse Clariom S Array PSR0100028874.mm.1 NM_025557.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028874.mm.1 NM_025557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028874.mm.1 TC0100003531.mm.1 MTA TC
          Mouse Expression Set 430 1452913_PM_at - -
          Mouse Expression Set 430 1452913_at - -
          Murine Genome U74v2 Set 97297_PM_at - -
          Murine Genome U74v2 Set 160264_s_at - -
          Murine Genome U74v2 Set 97297_at - -
          Murine Genome U74v2 Set 160264_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452913_at - -
          Mouse Genome 430A 2.0 Array 1452913_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452913_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452913_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 571903 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102480347 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218127 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254622 - -
          Illumina Sentrix mouse-6.v1 bead array 102480347 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102480347 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218127 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline