Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295411_g1
          See other ALPPL2 GE Assays ›
          Gene Symbol:
          Alppl2
          Gene Name:
          alkaline phosphatase, placental-like 2
          Gene Aliases:
          Akp5, C77216, D1Ertd816e, EAP
          Chromosome Location:
          Chr.1: 87086690 - 87096250 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295411_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alppl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007433.3 NP_031459.3 10 - 11 1461 54
          GenBank mRNA AK145571.1 - 11 - 12 1370 54
          AY054302.1 - 10 - 11 1302 54
          BC052874.1 - 10 - 11 1456 30033693 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002756.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alppl2

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          dephosphorylation
          catalytic activity
          alkaline phosphatase activity
          hydrolase activity
          phosphatase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20415724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20415726 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20415723 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 AY054302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 BC052874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 AK145571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 NM_007433.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 AY054302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 BC052874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 AK145571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132685 NM_007433.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766637 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140865 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 AY054302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 BC052874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 AK145571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 NM_007433.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 AY054302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 BC052874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 AK145571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132685 NM_007433.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 AY054302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 BC052874.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 AK145571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 NM_007433.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 AY054302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 BC052874.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 AK145571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132685 NM_007433.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20415656 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140865 - -
          Whole Mouse Genome G4122A A_51_P132685 AY054302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 BC052874.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 AK145571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 NM_007433.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 AY054302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 BC052874.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 AK145571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132685 NM_007433.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130857 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00440385 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00440384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00233864 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00440392 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00440393 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00233868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00440387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20415723 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20415724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20415726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20415648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00545803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00656422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00280564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00830633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00786788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00786791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00744335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00581763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00786787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00716904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01054360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00280565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01047235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00440393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01044071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00786792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20415639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00233890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00440387 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356365 - -
          Affymetrix HT MG-430 PM Array Plate 1450636_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1450635_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10356353 - -
          Mouse Genome 430 2.0 Array 1450635_at - -
          Mouse Genome 430 2.0 Array 1450636_s_at - -
          Mouse Genome 430 2.0 Array 1450636_PM_s_at - -
          Mouse Genome 430 2.0 Array 1450635_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11650_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11650_at - -
          Affymetrix Mouse Clariom S Array PSR0100022228.mm.1 NM_007433.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022228.mm.1 NM_007433.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022228.mm.1 TC0100002756.mm.1 MTA TC
          Mouse Expression Set 430 1450636_PM_s_at - -
          Mouse Expression Set 430 1450635_at - -
          Mouse Expression Set 430 1450636_s_at - -
          Mouse Expression Set 430 1450635_PM_at - -
          Murine Genome U74v2 Set 103991_at - -
          Murine Genome U74v2 Set 103991_PM_at - -
          Murine Genome U74v2 Set 102132_PM_i_at - -
          Murine Genome U74v2 Set 102132_i_at - -
          Mouse Genome 430A 2.0 Array 1450635_at - -
          Mouse Genome 430A 2.0 Array 1450636_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1450636_s_at - -
          Mouse Genome 430A 2.0 Array 1450635_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450635_at - -
          Affymetrix HT Mouse Genome 430A Array 1450635_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450636_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450636_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 563679 - -
          ABI Mouse Genome Survey Microarray 715168 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 50450 - -
          Illumina mouse-6 v1.0 expression beadchip 50450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666607 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666607 - -
          mouse-6 v1.1 (Illumina) scl16514.11.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 50450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16514.11.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2666607 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666607 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline