Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295761_m1
          See other LYRM7 GE Assays ›
          Gene Symbol:
          Lyrm7
          Gene Name:
          LYR motif containing 7
          Gene Aliases:
          1700024C24Rik, 9330147L21Rik
          Chromosome Location:
          Chr.11: 54826866 - 54860907 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295761_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyrm7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029327.3 NP_083603.2 4 - 5 269 81
          GenBank mRNA AK006294.1 - 4 - 5 595 81
          BC139403.1 - 4 - 5 400 8862425 81
          BC139406.1 - 4 - 5 400 8862428 81
          BY776521.1 - 4 - 5 269 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002769.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lyrm7

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812423 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812422 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812424 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812443 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593337 NM_029327.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593337 AK079070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593337 NM_029327.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593337 AK079070.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481980 AK006599.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481980 AK132722.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481980 AY550241.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481980 BC050810.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481980 NM_178386.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92758 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143004 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143208 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164566 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056986 AK006294.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593337 NM_029327.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593337 AK079070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593337 NM_029327.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593337 AK079070.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812424 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07348846 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07348902 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164566 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07348879 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02329606 - -
          Whole Mouse Genome G4122A A_51_P409751 BC139406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409751 BC139403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409751 AK006294.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143208 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164566 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02329606 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09221379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09221388 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09248859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09221377 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09018362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09221381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812423 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812424 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812422 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09580759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09580754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09221377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09221379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09562429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09670215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812406 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385939 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385936 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385938 - -
          Affymetrix HT MG-430 PM Array Plate 1430821_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385932 - -
          Mouse Genome 430 2.0 Array 1430821_PM_at - -
          Mouse Genome 430 2.0 Array 1430821_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75530_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75530_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100025067.mm.1 NM_029327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025067.mm.1 NM_029327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025067.mm.1 TC1100002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025072.mm.1 NM_029327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025072.mm.1 NM_029327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025072.mm.1 TC1100002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025070.mm.1 NM_029327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025070.mm.1 NM_029327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025070.mm.1 TC1100002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025071.mm.1 NM_029327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025071.mm.1 NM_029327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025071.mm.1 TC1100002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025073.mm.1 NM_029327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025073.mm.1 NM_029327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025073.mm.1 TC1100002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025069.mm.1 NM_029327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025069.mm.1 NM_029327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025069.mm.1 TC1100002769.mm.1 MTA TC
          Mouse Expression Set 430 1430821_PM_at - -
          Mouse Expression Set 430 1430821_at - -
          Murine Genome U74v2 Set 136452_at - -
          Murine Genome U74v2 Set 136452_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430821_at - -
          Affymetrix HT Mouse Genome 430B Array 1430821_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102450433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646036 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233917 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646036 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233917 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2500491 - -
          mouse-6 v1.1 (Illumina) scl0109286.1_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 102450433 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102450433 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2500491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0