Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295818_m1
          See other SERPINB13 GE Assays ›
          Gene Symbol:
          Serpinb13
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13
          Gene Aliases:
          5430417G24, HUR7, HURPIN, PI13, headpin
          Chromosome Location:
          Chr.1: 106980984 - 107001195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295818_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172852.3 NP_766440.2 7 - 8 825 71
          GenBank mRNA AK030670.1 - 7 - 8 827 71
          BC117990.1 - 7 - 8 815 40104505 71
          BC117991.1 - 7 - 8 762 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001026.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb13

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          negative regulation of keratinocyte apoptotic process
          protease binding
          serine-type endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P148902 NM_172852.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P148902 AK030670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P148902 NM_172852.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P148902 AK030670.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136640 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144374 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176076 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P148902 NM_172852.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P148902 AK030670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P148902 NM_172852.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P148902 AK030670.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511264 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176076 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04217303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144374 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04217272 - -
          Whole Mouse Genome G4122A A_51_P148902 NM_172852.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148902 AK030670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148902 NM_172852.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148902 AK030670.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110237 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176076 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00517890 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00269171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00884137 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00884136 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00517892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00273958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00221095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00517902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00273957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00269171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01107945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00741844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00382166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00273949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01107938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10349131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349137 - -
          Affymetrix Mouse Gene 1.1 ST Array 10349129 - -
          Affymetrix Mouse Clariom S Array PSR0100008500.mm.1 NM_172852.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008500.mm.1 NM_172852.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008500.mm.1 TC0100001026.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008501.mm.1 NM_172852.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008501.mm.1 NM_172852.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008501.mm.1 TC0100001026.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 749452 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4200072 - -
          Illumina mouse-6 v1.0 expression beadchip 4200072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256506 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256506 - -
          mouse-6 v1.1 (Illumina) scl17579.8.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17579.8.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256506 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256506 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline